Anti-RNA polymerase II Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: RNA polymerase II
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNBP1-49546
- NOVUNBP1-49546SS
- Antigen symbol:
- RNAP2
- RNAP2
- Antigen name:
- RNA polymerase 2
- RNA polymerase 2
- Antigen synonyms:
- RNA polymerase II subunit B1|hsRPB1|RPOL2|POLRA|RPBh1|RpIILS|DNA directed RNA polymerase II subunit RPB1|RP02|polymerase (RNA) II (DNA directed) polypeptide A|Polymerase (RNA) II (DNA directed) polypeptide A 220kDa|RNA polymerase II subunit 1|RPB220|DNA-directed RNA polymerase II largest subunit|RPO21|MGC75453|SUA8|220kDa|POLR2|hRPB220|POLR2A
- RNA polymerase II subunit B1|hsRPB1|RPOL2|POLRA|RPBh1|RpIILS|DNA directed RNA polymerase II subunit RPB1|RP02|polymerase (RNA) II (DNA directed) polypeptide A|Polymerase (RNA) II (DNA directed) polypeptide A 220kDa|RNA polymerase II subunit 1|RPB220|DNA-directed RNA polymerase II largest subunit|RPO21|MGC75453|SUA8|220kDa|POLR2|hRPB220|POLR2A
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 5430
- 5430
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice