Order Entry
Export
ContactUsLinkComponent
 
Anti-RBBP6 Rabbit Polyclonal Antibody (DyLight® 488)
  NOVUNBP1-49542G
Anti-RBBP6 Rabbit Polyclonal Antibody (DyLight® 488)
  NOVUNBP1-49542G

 

  • Antibody type:
    Primary
  • Antigen name:
    Retinoblastoma binding protein 6
  • Antigen symbol:
    RBBP6
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Gene ID:
    5930
  • Antigen synonyms:
    PACT|SNAMA|MY038|RBQ-1|P2P-R
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: RBBP6 (retinoblastoma binding protein 6)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human