Anti-C12orf5 Rabbit Polyclonal Antibody
The TIGAR / C12orf5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TIGAR / C12orf5. This antibody reacts with human, mouse. The TIGAR / C12orf5 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
There is 1 scientific publication using this antibody.
Type: Primary
Antigen: TIGAR/C12orf5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- NOVUNBP1-49534
- NOVUNBP1-49534SS
- Antigen symbol:
- TIGAR
- TIGAR
- Antigen name:
- TP53 Induced Glycolysis Regulatory Phosphatase
- TP53 Induced Glycolysis Regulatory Phosphatase
- Antigen synonyms:
- Chromosome 12 Open Reading Fame 5|C12orf5|FR2BP
- Chromosome 12 Open Reading Fame 5|C12orf5|FR2BP
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 57103
- 57103
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice