Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 550) [clone: exon 12]
Catalog # NOVUNBP1-47180R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:HIF1 alpha
- Antigen symbol:HIF1A
- Clonality:Polyclonal
- Clone:exon 12
- Conjugation:DyLight® 550
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:3091
- Antigen synonyms:PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone: exon 12
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human