Order Entry
Export
ContactUsLinkComponent
 
Anti-HIF1A Mouse Monoclonal Antibody (DyLight® 405) [clone: 162c]
  NOVUNBP1-30333V
Anti-HIF1A Mouse Monoclonal Antibody (DyLight® 405) [clone: 162c]
  NOVUNBP1-30333V

 

  • Antibody type:
    Primary
  • Antigen name:
    hypoxia inducible factor 1, alpha subunit inhibitor
  • Antigen symbol:
    HIF1AN
  • Clonality:
    Monoclonal
  • Clone:
    162c
  • Conjugation:
    DyLight® 405
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    55662
  • Antigen synonyms:
    FIH1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: 162c
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human