Anti-LOX propeptide Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-30327B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:LOX propeptide
- Clonality:Polyclonal
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:4015
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: LOX propeptide
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat