Order Entry
Export
ContactUsLinkComponent
Anti-FER Rabbit Polyclonal Antibody
Anti-FER Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-FER Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-20089SS
  • NOVUNBP1-20089
  • Antigen symbol:
  • FER
  • FER
  • Antigen name:
  • Tyrosine-protein kinase Fer
  • Tyrosine-protein kinase Fer
  • Antigen synonyms:
  • PPP1R74|p94-Fer|TYK3
  • PPP1R74|p94-Fer|TYK3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 2241
  • 2241
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information