Anti-HPi4 Mouse Monoclonal Antibody (DyLight® 488) [clone: HIC1-5F10]
Catalog # NOVUNBP1-18948G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Heterochromatin Protein 4
- Antigen symbol:HPI4
- Clonality:Monoclonal
- Clone:HIC1-5F10
- Conjugation:DyLight® 488
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: HPi4
Clonality: monoclonal
Clone: HIC1-5F10
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human