Order Entry
Export
ContactUsLinkComponent
 
Anti-MKI67 Rabbit Polyclonal Antibody (DyLight® 488)
  NOVUNB500-170G
Anti-MKI67 Rabbit Polyclonal Antibody (DyLight® 488)
  NOVUNB500-170G

 

  • Antibody type:
    Primary
  • Antigen name:
    Marker of proliferation Ki-67
  • Antigen symbol:
    MKI67
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Gene ID:
    4288
  • Antigen synonyms:
    MIB-1|MIB-|KIA|PPP1R105
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: MKI67 (marker of proliferation Ki-67)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: