Anti-ABCA7 Rat Monoclonal Antibody [clone: 7A1-144]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: ABCA7 (ATP-binding cassette, sub-family A (ABC1), member 7)
Clonality: monoclonal
Clone: 7A1-144
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Specifications
- Catalog No:
- NOVUNB400-166SS
- NOVUNB400-166
- Antigen symbol:
- ABCA7
- ABCA7
- Antigen name:
- ATP-binding cassette, Subfamily A (ABC1), Member 7
- ATP-binding cassette, Subfamily A (ABC1), Member 7
- Antigen synonyms:
- ATP-binding Cassette 1|CERP|ABC1|ABC Transporter 1|ABC 1|ATP-binding Cassette Transporter 1
- ATP-binding Cassette 1|CERP|ABC1|ABC Transporter 1|ABC 1|ATP-binding Cassette Transporter 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 7A1-144
- 7A1-144
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 10347
- 10347
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice