Anti-ABCA7 Rat Monoclonal Antibody (DyLight® 650) [clone: 7A1-144]
Catalog # NOVUNB400-163C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding cassette, Subfamily A (ABC1), Member 7
- Antigen symbol:ABCA7
- Clonality:Monoclonal
- Clone:7A1-144
- Conjugation:DyLight® 650
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:10347
- Antigen synonyms:ATP-binding Cassette 1|CERP|ABC1|ABC Transporter 1|ABC 1|ATP-binding Cassette Transporter 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ABCA7 (ATP-binding cassette, sub-family A (ABC1), member 7)
Clonality: monoclonal
Clone: 7A1-144
Conjugation: DyLight® 650
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse