Anti-CDKN2A Rat monoclonal antibody unconjugated [clone: 12-A1-1]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: monoclonal
Clone: 12-A1-1
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Specifications
- Catalog No:
- NOVUNB200-169SS
- NOVUNB200-169
- Antigen symbol:
- p19ARF
- p19ARF
- Antigen name:
- Cyclin-dependent kinase inhibitor 2A
- Cyclin-dependent kinase inhibitor 2A
- Antigen synonyms:
- CDKN2|p16-INK4|MLM|INK4|P16|p19ARF|p16-INK4a|p16INK4a|p16|CDKN2A|p16INK4|p19|p14|p16INK4A|CDK4I|INK4a|p14ARF|CMM2|ARF|MTS1|TP16
- CDKN2|p16-INK4|MLM|INK4|P16|p19ARF|p16-INK4a|p16INK4a|p16|CDKN2A|p16INK4|p19|p14|p16INK4A|CDK4I|INK4a|p14ARF|CMM2|ARF|MTS1|TP16
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 12-A1-1
- 12-A1-1
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 12578
- 12578
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice