Anti-AQP2 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The Aquaporin-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Aquaporin-2. This antibody reacts with human, mouse, rat. The Aquaporin-2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:200
Type: Primary
Antigen: Aquaporin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- NOVUNB110-74682
- NOVUNB110-74682SS
- Antigen symbol:
- AQP2
- AQP2
- Antigen name:
- Aquaporin 2
- Aquaporin 2
- Antigen synonyms:
- WCH-CD|water channel protein for renal collecting duct|water-channel aquaporin 2|AQP-2|aquaporin-CD|collecting duct water channel protein|aquaporin-2|AQP-CD|ADH water channel
- WCH-CD|water channel protein for renal collecting duct|water-channel aquaporin 2|AQP-2|aquaporin-CD|collecting duct water channel protein|aquaporin-2|AQP-CD|ADH water channel
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 359
- 359
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice