Anti-TERF1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 57-6]
Catalog # NOVUNB110-68281H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Telomeric Repeat Binding Factor 1
- Antigen symbol:TERF1
- Clonality:Monoclonal
- Clone:57-6
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:7013
- Antigen synonyms:TERF1|telomeric repeat binding factor (NIMA-interacting) 1|TRF1|hTRF1-AS|TRBF1|PIN2|FLJ41416|t-TRF1|TRF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: TERF1 (telomeric repeat binding factor (NIMA-interacting) 1)
Clonality: monoclonal
Clone: 57-6
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: