Anti-TERF1 Mouse Monoclonal Antibody [clone: 57-6]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: TERF1 (telomeric repeat binding factor (NIMA-interacting) 1)
Clonality: monoclonal
Clone: 57-6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Specifications
- Catalog No:
- NOVUNB110-68281
- NOVUNB110-68281SS
- Antigen symbol:
- TERF1
- TERF1
- Antigen name:
- Telomeric Repeat Binding Factor 1
- Telomeric Repeat Binding Factor 1
- Antigen synonyms:
- TERF1|telomeric repeat binding factor (NIMA-interacting) 1|TRF1|hTRF1-AS|TRBF1|PIN2|FLJ41416|t-TRF1|TRF
- TERF1|telomeric repeat binding factor (NIMA-interacting) 1|TRF1|hTRF1-AS|TRBF1|PIN2|FLJ41416|t-TRF1|TRF
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 57-6
- 57-6
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 7013
- 7013
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice