Anti-RB1 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56888
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Retinoblastoma 1
- Antigen symbol:RB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:5925
- Antigen synonyms:pRb|OSRC|pp110|PPP1R130|RB|p105-Rb
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 μL
- Pk:0,2 mL
Specifications
About this item
Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: