Anti-TEM8/ANTXR1 Mouse Monoclonal Antibody [clone: 200C1339(SB20)]
NOVUNB100-56585
- Antibody type:Primary
- Antigen name:Anthrax Toxin Receptor 1
- Antigen symbol:ANTXR1
- Clonality:Monoclonal
- Clone:200C1339(SB20)
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:84168
- Antigen synonyms:ATR|TEM8|FLJ21776|FLJ10601|FLJ11298
- Storage buffer:PBS containing 0,05% BSA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
The TEM8 / ANTXR1 Antibody (200C1339(SB20)) from Novus Biologicals is a mouse monoclonal antibody to TEM8 / ANTXR1. This antibody reacts with human. The TEM8 / ANTXR1 Antibody (200C1339(SB20)) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 25 ?g/ml
Type: Primary
Antigen: TEM8/ANTXR1
Clonality: Monoclonal
Clone: 200C1339(SB20)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human