Anti-Alas1 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56415
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Aminolevulinate, delta-, Synthase 1
- Antigen symbol:Alas1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:211
- Antigen synonyms:Alas1
- Storage buffer:PBS containing 0,05% BSA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
The Alas1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Alas1. This antibody reacts with human. The Alas1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry-Paraffin.
Simple Western,
Type: Primary
Antigen: Alas1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human