Order Entry
Export
ContactUsLinkComponent
Anti-INHAT-1 Rabbit Polyclonal Antibody
Anti-INHAT-1 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56353
Supplier:  Novus Biologicals
undefined
Anti-INHAT-1 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56353
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    INHAT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: INHAT-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat