Anti-TRAF-2 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56173
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:TNF receptor-associated factor 2
- Antigen symbol:TRAF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:7186
- Antigen synonyms:TRAP|TRAP3|TNF receptor-associated factor 2|TRAF2|MGC,45012|E3 ubiquitin-protein ligase TRAF2
- Storage buffer:Whole antisera
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
The TRAF-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRAF-2. This antibody reacts with human. The TRAF-2 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000
Type: Primary
Antigen: TRAF-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human