Anti-OS9 Rabbit Polyclonal Antibody (Biotin)
NOVUNB100-519B
- Antibody type:Primary
- Antigen name:Osteosarcoma amplified 9, endoplasmic reticulum lectin
- Antigen symbol:OS9
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:10956
- Antigen synonyms:ERLEC2|OS-9
- Storage buffer:PBS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:0,1 mL
The OS9 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to OS9. This antibody reacts with human, mouse. The OS9 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:250
Type: Primary
Antigen: OS9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse