Anti-Speedy/Ringo Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB100-2521F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:speedy/RINGO cell cycle regulator family member A
- Antigen symbol:SPDYA
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Western blot:Yes
- Gene ID:245711
- Antigen synonyms:RINGOA|SPDY1|RINGO3|SPY1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Speedy/Ringo
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: