Order Entry
Export
ContactUsLinkComponent
 
Anti-ZMPSTE24 Rabbit Polyclonal Antibody
  NOVUNB100-2388
undefined
Anti-ZMPSTE24 Rabbit Polyclonal Antibody
  NOVUNB100-2388

 

  • Antibody type:
    Primary
  • Antigen name:
    Zinc metallopeptidase STE24
  • Antigen symbol:
    ZMPSTE24
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    10269
  • Antigen synonyms:
    PRO1|STE24|Ste24p|FACE1|FACE-1|HGPS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: ZMPSTE24 (zinc metallopeptidase STE24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human