Anti-SEMA3B Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNB100-2218C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Semaphorin 3B
- Antigen symbol:SEMA3B
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:7869
- Antigen synonyms:SEMA5|SEMAA|LUCA-1|SemA|semaV
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SEMA3B (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: