Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: B486A1]
NOVUNB100-2181APC
- Antibody type:Primary
- Antigen name:CD4 molecule
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:B486A1
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:920
- Antigen synonyms:CD4mut
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: B486A1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human