Order Entry
Export
ContactUsLinkComponent
Anti-CNX Rabbit Polyclonal Antibody
Anti-CNX Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
Anti-CNX Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-1974-B
  • NOVUNB100-1974-A
  • Antigen symbol:
  • CANX
  • CANX
  • Antigen name:
  • calnexin
  • calnexin
  • Antigen synonyms:
  • CNX|P90|IP90
  • CNX|P90|IP90
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 821
  • 821
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage buffer:
  • PBS and 30% Glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications