Order Entry
Export
ContactUsLinkComponent
Anti-HIF1A Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: H1alpha67]
Anti-HIF1A Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: H1alpha67]
Catalog # NOVUNB100-123PCP
Supplier:  Novus Biologicals
Anti-HIF1A Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: H1alpha67]
Catalog # NOVUNB100-123PCP
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor)
  • Antigen symbol:
    HIF1A
  • Clonality:
    Monoclonal
  • Clone:
    H1alpha67
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Gene ID:
    3091
  • Antigen synonyms:
    PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: H1alpha67
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: