Anti-OS9 Rabbit Polyclonal Antibody
Type: Primary
Antigen: OS9 (osteosarcoma amplified 9, endoplasmic reticulum lectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
- Catalog No:
- NOVUBC100-520
- NOVUBC100-520SS
- Antigen symbol:
- OS9
- OS9
- Antigen name:
- Osteosarcoma amplified 9, endoplasmic reticulum lectin
- Osteosarcoma amplified 9, endoplasmic reticulum lectin
- Antigen synonyms:
- ERLEC2|OS-9
- ERLEC2|OS-9
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 10956
- 10956
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice