Order Entry
Export
ContactUsLinkComponent
Anti-SLC22A7 Rabbit Polyclonal Antibody
Anti-SLC22A7 Rabbit Polyclonal Antibody
Catalog # BSBTPA2190
Supplier:  Boster Bio
undefined
Anti-SLC22A7 Rabbit Polyclonal Antibody
Catalog # BSBTPA2190
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 22 Member A7
  • Antigen symbol:
    Solute carrier family 22, member 7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    OAT2|NLT
  • Amino acid number:
    514 to 530
  • Sequence:
    ETRQAQLPETIQDVERK
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human SLC22A7 (514-530aa ETRQAQLPETIQDVERK), different from the related rat and mouse sequences by two amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: SLC22A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat