Order Entry
Export
ContactUsLinkComponent
Anti-CCR3 Rabbit Polyclonal Antibody
Anti-CCR3 Rabbit Polyclonal Antibody
Catalog # BSBTPA2176
Supplier:  Boster Bio
undefined
Anti-CCR3 Rabbit Polyclonal Antibody
Catalog # BSBTPA2176
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chemokine (C-C motif) receptor 3
  • Antigen symbol:
    CCR3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    CD193|eotaxin receptor|MIP1-alpha receptor like-2|CC CKR3
  • Amino acid number:
    1 - 20
  • Sequence:
    MTTSLDTVETFGTTSYYDDV
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human CCR3 (1-20aa MTTSLDTVETFGTTSYYDDV)
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for C-C chemokine receptor type 3(CCR3) detection. Tested with WB, IHC-P, ICC in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human