Order Entry
Export
ContactUsLinkComponent
Anti-Prolactin Receptor Rabbit Polyclonal Antibody
Anti-Prolactin Receptor Rabbit Polyclonal Antibody
Catalog # BSBTPA2087
Supplier:  Boster Bio
undefined
Anti-Prolactin Receptor Rabbit Polyclonal Antibody
Catalog # BSBTPA2087
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Prolactin Receptor
  • Antigen symbol:
    PRL-R
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    secreted prolactin binding protein|delta 4-delta 7/11 truncated prolactin receptor|prolactin receptor delta 7/11|hPRL receptor|Delta 4-SF1b truncated prolactin receptor
  • Amino acid number:
    591 to 605
  • UniProtKB:
    Q08501
  • Sequence:
    QLGRLDYLDPTCFMH
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of mouse Prolactin Receptor (591-605aa QLGRLDYLDPTCFMH), identical to the related rat sequence, and different from the related human sequence by three amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Prolactin receptor(PRLR) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

PRLR(Prolactin Receptor), is a cytokine receptor. By somatic cell hybrid analysis and by in situ hybridization, Arden et al.(1989, 1990) demonstrated that the prolactin receptor gene resides in the same chromosomal region as the growth hormone receptor gene, which has been mapped to 5p13-p12. Cunningham et al.(1990) demonstrated that zinc greatly increases the affinity of GH for the extracellular binding domain of PRLR, although it is not required for binding of GH to the growth hormone receptor or for binding of prolactin to the prolactin receptor. By mutational analysis, they showed that a cluster of 3 residues(histidine-18, histidine-21, and glutamic acid-174) in GH and histidine-188 in PRLR(conserved in all PRL receptors but not GH receptors) are likely zinc-ion ligands.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: Prolactin Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat