Anti-SPHK1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1996
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Sphingosine Kinase 1
- Antigen symbol:SPHK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:sphingosine kinase 1|SPHK|SK 1|SK1|SPK|SPK1.|SPHK 1|SPK 1
- Amino acid number:117 to 133
- Sequence:AASLNHYAGYEQVTNED
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human SPHK1 (117-133aa AASLNHYAGYEQVTNED), different from the related rat sequence by two amino acids, and from the related mouse sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SPHK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse