Anti-OSBP1 Rabbit Polyclonal Antibody
BSBTPA1949
- Antibody type:Primary
- Antigen name:Oxysterol Binding Protein 1
- Antigen symbol:OSBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:OSBP1|OSBP|oxysterol binding protein
- Amino acid number:290 - 304
- Sequence:WQKSLQYERDQRIRL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human OSBP1 (290-304aa WQKSLQYERDQRIRL), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Oxysterol-binding protein 1 (OSBP) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: OSBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat