Order Entry
Export
ContactUsLinkComponent
Anti-ErbB 4 Rabbit Polyclonal Antibody
Anti-ErbB 4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1881
Supplier:  Boster Bio
undefined
Anti-ErbB 4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1881
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Erb-b2 receptor tyrosine kinase 4
  • Antigen symbol:
    ERBB4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    ALS19|p180erbB4|HER4
  • Amino acid number:
    35 to 51
  • UniProtKB:
    Q15303
  • Sequence:
    KLSSLSDLEQQYRALRK
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human ErbB 4 (35-51aa KLSSLSDLEQQYRALRK), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Receptor tyrosine-protein kinase erbB-4(ERBB4) detection. Tested with WB in Human;Mouse;Rat.

ERBB4(V-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4) also known as ONCOGENE ERBB4 or HER4, is an enzyme that in humans is encoded by the ERBB4 gene. The HER4/ERBB4 gene is a member of the type I receptor tyrosine kinase subfamily that includes EGFR, ERBB2 and ERBB3. The ERBB4 gene is mapped on 2q34. ERBB4 is a single-pass type I transmembrane protein with multiple furin-like cysteine rich domains, a tyrosine kinase domain, a phosphotidylinositol-3 kinase binding site and a PDZ domainbinding motif. Huang et al.(2000) reported that ERBB4 is enriched in the postsynaptic density and associates with PSD95. ERBB4 is a transmembrane receptor tyrosine kinase that regulates cell proliferation and differentiation. After binding its ligand, heregulin, or activation of protein kinase C by TPA, the ERBB4 ectodomain is cleaved by a metalloprotease. Ni et al.(2001)concluded that gamma-secretase cleavage of ERBB4 may represent another mechanism for receptor tyrosine kinase-mediated signaling.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

WB: The detection limit for ERBB4 is approximately 0.5ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: ErbB 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat