Order Entry
Export
ContactUsLinkComponent
Anti-LAMP1 Rabbit Polyclonal Antibody
Anti-LAMP1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1822
Supplier:  Boster Bio
undefined
Anti-LAMP1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1822
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Lysosomal-associated membrane protein 1
  • Antigen symbol:
    LAMP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    LGP120|CD107a|CD107 antigen-like family member A|Lysosome-associated membrane glycoprotein 1|LAMP-1|LAMPA
  • Amino acid number:
    403 - 417
  • Sequence:
    YLVGRKRSHAGYQTI
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human LAMP1 (403-417aa YLVGRKRSHAGYQTI), different from the related mouse and rat sequences by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Lysosome-associated membrane glycoprotein 1 (LAMP1) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: LAMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat