Anti-VDAC/Porin Rabbit Polyclonal Antibody
Catalog # BSBTPA1780
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Voltage-dependent anion-selective channel protein 1
- Antigen symbol:VDAC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:Voltage-dependent anion-selective channel protein 1|PORIN-31-HL|Plasmalemmal porin|Outer mitochondrial membrane protein porin|PORIN|MGC111064|VD
- Amino acid number:163 - 178
- Sequence:RVTQSNFAVGYKTDEF
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human VDAC (163-178aa RVTQSNFAVGYKTDEF), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Voltage-dependent anion-selective channel protein 1 (VDAC1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: VDAC/Porin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat