Order Entry
Export
ContactUsLinkComponent
Anti-MCM6 Rabbit Polyclonal Antibody
Anti-MCM6 Rabbit Polyclonal Antibody
Catalog # BSBTPA1769
Supplier:  Boster Bio
undefined
Anti-MCM6 Rabbit Polyclonal Antibody
Catalog # BSBTPA1769
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Minichromosome Maintenance Complex Component 6
  • Antigen symbol:
    MCM6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    MCG40308|Mis5|minichromosome maintenance complex component 6|P105MCM
  • Amino acid number:
    589 to 605
  • UniProtKB:
    Q14566
  • Sequence:
    ESEDFIVEQYKHLRQRD
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human MCM6 (589-605aa ESEDFIVEQYKHLRQRD), different from the related rat and mouse sequences by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for DNA replication licensing factor MCM6(MCM6) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

MCM6(Minichromosome maintenance, s. pombe, homolog of, 6) is a protein that in humans is encoded by the MCM6 gene. MCM6 is one of the highly conserved mini-chromosome maintenance proteins(MCM) that are essential for the initiation of eukaryotic genome replication. The MCM genes were originally identified in yeast defective in minichromosome maintenance and have since been shown to play roles in the progression of the cell cycle; many are cell division control genes. The MCM6 gene is mapped on 2q21.3. Mcm 6 has recently been shown to interact strongly Cdt1 at defined residues, by mutating these target residues Wei et al. observed lack of Cdt1 recruitment of Mcm2-7 to the pre-RC. An approximately 200-kb region surrounding the C/T(-13910) polymorphism in MCM6 intron 13 functioned as an enhancer of the lactase gene promoter in intestinal cell culture.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: MCM6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat