Anti-Kallikrein 6 Rabbit Polyclonal Antibody
Catalog # BSBTPA1767
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Kallikrein 6
- Antigen symbol:KLK6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:PRSS18|KLK6|Kallikrein-6|PRSS9|Neurosin|Zyme|Serine protease 18|SP59|Protease M|Serine protease 9
- Amino acid number:227 - 244
- Sequence:YTNVCRYTNWIQKTIQAK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Kallikrein 6 (227-244aa YTNVCRYTNWIQKTIQAK).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Kallikrein-6 (KLK6) detection. Tested with WB, IHC-P, ICC in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Kallikrein 6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human