Order Entry
Export
ContactUsLinkComponent
 
Anti-TRPC3 Rabbit Polyclonal Antibody
  BSBTPA1753
undefined
Anti-TRPC3 Rabbit Polyclonal Antibody
  BSBTPA1753

 

  • Antibody type:
    Primary
  • Antigen name:
    Transient Receptor Potential Cation Channel, subfamily C, member 3
  • Antigen symbol:
    TRPC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    TRPC3|Transient receptor protein 3|TrpC3|TRP3|Short transient receptor potential channel 3
  • Amino acid number:
    836 - 851
  • Sequence:
    HSFNSILNQPTRYQQI
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human TRPC3 (836-851aa HSFNSILNQPTRYQQI), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Short transient receptor potential channel 3 (TRPC3) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: TRPC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat