Anti-Serotonin transporter Rabbit Polyclonal Antibody
BSBTPA1705
- Antibody type:Primary
- Antigen name:Serotonin Transporter
- Antigen symbol:5-HTT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:HTT|5HTT|5-HTT |SERT|hSERT|OCD1
- Amino acid number:2 - 24
- Sequence:NSQKVLSECKDREDCQEN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Serotonin transporter (7-24aa NSQKVLSECKDREDCQEN).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Sodium-dependent serotonin transporter (SLC6A4) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Serotonin transporter
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human