Order Entry
Export
ContactUsLinkComponent
 
Anti-Chk2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1655
Supplier:  Boster Bio
undefined
Anti-Chk2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1655
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Checkpoint Kinase 2
  • Antigen symbol:
    CHK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    PP1425|LFS2|RAD53|CDS1|OTTHUMP00000028871|bA444G7|HuCds1|CHK2|Cds1
  • Amino acid number:
    133 to 148
  • Sequence:
    TDKYRTYSKKHFRIFR
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human Chk2 (133-148aa TDKYRTYSKKHFRIFR), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: Chk2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat