Anti-Kallikrein 2 Rabbit Polyclonal Antibody
BSBTPA1626
- Antibody type:Primary
- Antigen name:Kallikrein 2
- Antigen symbol:KLK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Glandular kallikrein-1|Tissue kallikrein 2|hGK-1
- Amino acid number:243 - 261
- Sequence:YTKVVHYRKWIKDTIAANP
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Kallikrein 2 (243-261aa YTKVVHYRKWIKDTIAANP).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Kallikrein-2(KLK2) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Kallikrein 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human