Order Entry
Export
ContactUsLinkComponent
Anti-IL2 Receptor gamma Rabbit Polyclonal Antibody
Anti-IL2 Receptor gamma Rabbit Polyclonal Antibody
Catalog # BSBTPA1616
Supplier:  Boster Bio
undefined
Anti-IL2 Receptor gamma Rabbit Polyclonal Antibody
Catalog # BSBTPA1616
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 2 Receptor Gamma
  • Antigen symbol:
    IL2RG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    IL2 Receptor gamma|CD132|CIDX|IL-2RG|SCIDX1|IMD4|P64|SCIDX
  • Amino acid number:
    88 - 102
  • Sequence:
    HYWYKNSDNDKVQKC
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human IL2 Receptor gamma (88-102aa HYWYKNSDNDKVQKC).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Cytokine receptor common subunit gamma(IL2RG) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: IL2 Receptor gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human