Order Entry
Export
ContactUsLinkComponent
Anti-SPARC Rabbit Polyclonal Antibody
Anti-SPARC Rabbit Polyclonal Antibody
Catalog # BSBTPA1585
Supplier:  Boster Bio
undefined
Anti-SPARC Rabbit Polyclonal Antibody
Catalog # BSBTPA1585
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Secreted Protein, Acidic, Cysteine-Rich
  • Antigen symbol:
    SPARC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    Cysteine-Rich Protein|Secreted Protein Acidic And Rich In Cysteine|ON|Basement-Membrane Protein 40|Osteonectin|BM-40
  • Amino acid number:
    284 - 303
  • Sequence:
    DEWAGCFGIKQKDIDKDLVI
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human SPARC (284-303aa DEWAGCFGIKQKDIDKDLVI).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for SPARC(SPARC) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human