Anti-IL7 Rabbit Polyclonal Antibody
BSBTPA1467
- Antibody type:Primary
- Antigen name:Interleukin 7
- Antigen symbol:IL7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:IL-7|IL7|Interluekin-7|Interluekin 7|IL 7|interluekin
- Amino acid number:26 - 42
- Sequence:ECHIKDKEGKAYESVLM
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of mouse IL7 (26-42aa ECHIKDKEGKAYESVLM).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Interleukin-7(IL7) detection. Tested with WB, IHC-P in Mouse.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: IL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse