Order Entry
Export
ContactUsLinkComponent
Anti-Caspase-1(P20) Rabbit Polyclonal Antibody
Anti-Caspase-1(P20) Rabbit Polyclonal Antibody
  BSBTPA1440-1
undefined
Anti-Caspase-1(P20) Rabbit Polyclonal Antibody
  BSBTPA1440-1

 

  • Antibody type:
    Primary
  • Antigen name:
    Caspase 1
  • Antigen symbol:
    CASP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    137 - 157
  • Sequence:
    LEKAQKLWKENPSEIYPIMNT
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of mouse CASP1 (137-157aa LEKAQKLWKENPSEIYPIMNT), different from the related rat sequence by three amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Caspase-1(CASP1) detection. Tested with WB in Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Caspase-1(P20)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat