Anti-Calpain 1 Rabbit Polyclonal Antibody
BSBTPA1364
- Antibody type:Primary
- Antigen name:Calpain 1
- Antigen symbol:CAPN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:muCANP|CANP|CANPL1|CANP1|muCL
- Amino acid number:312 - 326
- Sequence:EWNNVDPYERDQLRV
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Calpain 1 (312-326aa EWNNVDPYERDQLRV), different from the mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Calpain-1 catalytic subunit(CAPN1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Calpain 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat