Anti-IL-18 Rabbit Polyclonal Antibody
Catalog # BSBTPA1342
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 18
- Antigen symbol:IL18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:IL-1g|IL-1 gamma|interleukin 18 (interferon-gamma-inducing factor)|interleukin-1 gamma|interleukin-18|interferon-gamma-inducing factor|MGC12320|interleukin 18|IL1F4|IGIF|IL18|IL-18
- Amino acid number:175 - 193
- Sequence:KKEDELGDRSIMFTVQNED
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of Human IL18 (175-193aa KKEDELGDRSIMFTVQNED)
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Interleukin-18 (IL18) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: IL-18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human