Anti-Caspase-3(P10) Rabbit Polyclonal Antibody
Catalog # BSBTPA1302-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Caspase 3
- Antigen symbol:CASP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:procaspase3|Yama|CASP-3|CASP3|CPP32|Apopain|CPP32B|SCA-1|CPP-32|apopain
- Amino acid number:220 - 236
- Sequence:CAMLKQYADKLEFMHIL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Caspase-3 (P10) (220-236aa CAMLKQYADKLEFMHIL)
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Caspase-3 (CASP3) detection. Tested with WB, IHC-P, ICC in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Caspase-3(P10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human