Anti-ROCK2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1242
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Rho-associated, Coiled-coil-containing Protein Kinase 2
- Antigen symbol:ROCK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:Rho kinase 2|ROCK-II|Rho-associated|p164 ROCK-2|Rho-associated protein kinase 2|coiled-coil-containing protein kinase 2|KIAA619|coiled-coil-containing protein kinase II
- Amino acid number:35 - 51
- Sequence:RDPRSPINVESLLDGLN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human ROCK2 (35-51aa RDPRSPINVESLLDGLN), identical to the related rat and mouse sequences
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Rho-associated protein kinase 2 (ROCK2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: ROCK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat